The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006092.2 | NP_006083.1 | 5 - 6 | 899 | 59 |
XM_005249568.1 | XP_005249625.1 | 6 - 7 | 954 | 59 | |
XM_005249572.1 | XP_005249629.1 | 4 - 5 | 817 | 59 | |
XM_006715633.2 | XP_006715696.1 | 6 - 7 | 991 | 59 | |
XM_011515079.1 | XP_011513381.1 | 7 - 8 | 1137 | 59 | |
XM_011515080.2 | XP_011513382.1 | 5 - 6 | 813 | 59 | |
XM_011515081.2 | XP_011513383.1 | 5 - 6 | 963 | 59 | |
XM_011515083.1 | XP_011513385.1 | 6 - 7 | 954 | 59 | |
XM_011515084.1 | XP_011513386.1 | 6 - 7 | 954 | 59 | |
XM_011515085.1 | XP_011513387.1 | 6 - 7 | 954 | 59 | |
XM_011515087.1 | XP_011513389.1 | 6 - 7 | 954 | 59 | |
XM_011515088.2 | XP_011513390.1 | 6 - 7 | 954 | 59 | |
XM_017011674.1 | XP_016867163.1 | 6 - 7 | 954 | 59 | |
XM_017011675.1 | XP_016867164.1 | 5 - 6 | 860 | 59 | |
GenBank mRNA | AF113925.1 | - | 5 - 6 | 800 | 59 |
AF126484.1 | - | 5 - 6 | 827 | 59 | |
AK023969.1 | - | 5 - 6 | 860 | 59 | |
AK300367.1 | - | 5 - 6 | 818 | 59 | |
BC040339.1 | - | 5 - 6 | 814 | 59 | |
KU178514.1 | - | 2 - 3 | 376 | 59 |