The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006158.4 | NP_006149.2 | 1 - 2 | 1399 | 71 |
GenBank mRNA | AK057731.1 | - | 2 - 3 | 973 | 71 |
AK075003.1 | - | 1 - 2 | 1142 | 71 | |
AK225975.1 | - | 1 - 2 | 1142 | 71 | |
AK299095.1 | - | 2 - 3 | 984 | 71 | |
AK314591.1 | - | 1 - 2 | 1142 | 71 | |
AL713644.1 | - | 2 - 3 | 575 | 71 | |
AY156690.1 | - | 1 - 2 | 1045 | 71 | |
BC039237.1 | - | 1 - 2 | 1142 | 71 | |
KU178228.1 | - | 2 - 3 | 766 | 71 | |
KU178229.1 | - | 1 - 2 | 1045 | 71 |