The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006226.3 | NP_006217.3 | 5 - 6 | 3508 | 65 |
XM_005246643.3 | XP_005246700.1 | 5 - 6 | 2935 | 65 | |
XM_005246644.3 | XP_005246701.1 | 5 - 6 | 2888 | 65 | |
XM_011511351.2 | XP_011509653.1 | 6 - 7 | 3565 | 65 | |
XM_017004339.1 | XP_016859828.1 | 5 - 6 | 2982 | 65 | |
XM_017004340.1 | XP_016859829.1 | 5 - 6 | 3729 | 65 | |
GenBank mRNA | AK127514.1 | - | 6 - 7 | 3565 | 65 |
AK302673.1 | - | 5 - 6 | 2935 | 65 | |
BC101531.1 | - | 4 - 5 | 2852 | 65 | |
BC111985.1 | - | 4 - 5 | 2852 | 65 | |
BX537442.1 | - | 4 - 5 | 890 | 65 | |
D42108.1 | - | 5 - 6 | 3019 | 65 |