The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281463.1 | NP_001268392.1 | 16 - 17 | 2692 | 63 |
NM_006306.3 | NP_006297.2 | 15 - 16 | 2546 | 63 | |
GenBank mRNA | AK291921.1 | - | 15 - 16 | 2505 | 63 |
BC064368.1 | - | 15 - 16 | 2477 | 63 | |
BC112127.1 | - | 15 - 16 | 2486 | 63 | |
BC171726.1 | - | 16 - 17 | 2632 | 63 | |
BX640922.1 | - | 15 - 16 | 2589 | 63 | |
D80000.2 | - | 15 - 16 | 2418 | 63 | |
S78271.1 | - | 15 - 16 | 2451 | 63 |