The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042734.3 | NP_001036199.1 | 17 - 18 | 3125 | 76 |
NM_001300813.2 | NP_001287742.1 | 19 - 20 | 3320 | 76 | |
NM_001318085.1 | NP_001305014.1 | 18 - 19 | 3227 | 76 | |
NM_001318086.1 | NP_001305015.1 | 17 - 18 | 3122 | 76 | |
NM_006323.4 | NP_006314.2 | 18 - 19 | 3230 | 76 | |
XM_005262688.1 | XP_005262745.1 | 19 - 20 | 3199 | 76 | |
XM_005262691.1 | XP_005262748.1 | 18 - 19 | 3091 | 76 | |
XM_011531535.2 | XP_011529837.1 | 18 - 19 | 3094 | 76 | |
XM_011531536.2 | XP_011529838.1 | 18 - 19 | 3303 | 76 | |
XM_011531537.2 | XP_011529839.1 | 18 - 19 | 3009 | 76 | |
XM_011531539.2 | XP_011529841.1 | 17 - 18 | 2911 | 76 | |
XM_011531540.2 | XP_011529842.1 | 13 - 14 | 1740 | 76 | |
XM_017007652.1 | XP_016863141.1 | 16 - 17 | 2844 | 76 | |
GenBank mRNA | AJ131245.1 | - | 18 - 19 | 3230 | 76 |
AK300278.1 | - | 17 - 18 | 2913 | 76 | |
AK304234.1 | - | 15 - 16 | 1970 | 76 | |
BC040137.1 | - | 18 - 19 | 3088 | 76 | |
BC117135.1 | - | 17 - 18 | 3068 | 76 | |
BC143268.1 | - | 19 - 20 | 3263 | 76 | |
BC143276.1 | - | 17 - 18 | 3068 | 76 |