The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006392.3 | NP_006383.2 | 3 - 4 | 333 | 62 |
NR_027700.2 | 3 - 4 | 333 | 62 | ||
GenBank mRNA | AK292721.1 | - | 3 - 4 | 233 | 62 |
BC018421.1 | - | 2 - 3 | 193 | 62 | |
BC035369.1 | - | 3 - 4 | 223 | 62 | |
BC104791.1 | - | 3 - 4 | 333 | 62 | |
BC104793.1 | - | 3 - 4 | 333 | 62 | |
BX647439.1 | - | 3 - 4 | 233 | 62 | |
BX647513.1 | - | 2 - 3 | 257 | 62 | |
DA805367.1 | - | 3 - 4 | 278 | 62 | |
Y12065.1 | - | 2 - 3 | 243 | 62 |