Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATG7
Entrez Gene ID
10533
Gene Name
autophagy related 7
Gene Aliases
APG7-LIKE, APG7L, GSA7
Chromosome Location
Chr.3: 11272324 - 11564704 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136031.2 NP_001129503.2 11 - 12 1243 77
NM_001144912.1 NP_001138384.1 10 - 11 1126 77
NM_006395.2 NP_006386.1 11 - 12 1243 77
XM_006712931.3 XP_006712994.1 11 - 12 1186 77
XM_006712932.3 XP_006712995.1 12 - 13 1450 77
XM_006712933.3 XP_006712996.1 11 - 12 1423 77
XM_011533277.2 XP_011531579.1 13 - 14 1574 77
XM_011533278.2 XP_011531580.1 12 - 13 1279 77
XM_011533279.2 XP_011531581.1 13 - 14 1574 77
XM_011533280.2 XP_011531582.1 13 - 14 1574 77
XM_011533281.2 XP_011531583.1 13 - 14 1574 77
XM_011533282.2 XP_011531584.1 13 - 14 1574 77
XM_011533283.2 XP_011531585.1 13 - 14 1574 77
XM_011533284.2 XP_011531586.1 13 - 14 1574 77
XM_011533286.2 XP_011531588.1 13 - 14 1574 77
XM_017005542.1 XP_016861031.1 11 - 12 1160 77
XM_017005543.1 XP_016861032.1 11 - 12 1186 77
XM_017005544.1 XP_016861033.1 12 - 13 1448 77
XM_017005545.1 XP_016861034.1 11 - 12 1228 77
XM_017005546.1 XP_016861035.1 12 - 13 1279 77
XM_017005547.1 XP_016861036.1 13 - 14 1574 77
XM_017005548.1 XP_016861037.1 11 - 12 1186 77
XM_017005551.1 XP_016861040.1 11 - 12 1186 77
XM_017005553.1 XP_016861042.1 8 - 9 816 77
XM_017005554.1 XP_016861043.1 8 - 9 816 77
GenBank mRNA AF094516.1 - 10 - 11 1151 77
AK075221.1 - 11 - 12 1170 77
AK298833.1 - 11 - 12 1121 77
AK303694.1 - 10 - 11 1053 77
AK314504.1 - 11 - 12 1160 77
AL122075.1 - 5 - 6 592 77
BC000091.2 - 11 - 12 1158 77

View Full Product Details