The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006398.3 | NP_006389.2 | 1 - 2 | 250 | 67 |
GenBank mRNA | AF123050.1 | - | 1 - 2 | 250 | 67 |
AK311914.1 | - | 1 - 2 | 57 | 67 | |
BC012472.1 | - | 1 - 2 | 50 | 67 | |
DB181897.1 | - | 1 - 2 | 161 | 67 | |
Y12653.1 | - | 1 - 2 | 44 | 67 |