The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006421.4 | NP_006412.2 | 34 - 35 | 5211 | 81 |
XM_005251134.4 | XP_005251191.1 | 35 - 36 | 5190 | 81 | |
XM_005251135.3 | XP_005251192.1 | 34 - 35 | 5443 | 81 | |
XM_005251136.3 | XP_005251193.1 | 34 - 35 | 5461 | 81 | |
XM_011517442.2 | XP_011515744.1 | 35 - 36 | 5497 | 81 | |
GenBank mRNA | AB209324.1 | - | 24 - 25 | 3256 | 81 |
AF084520.1 | - | 34 - 35 | 4821 | 81 | |
AF111162.1 | - | 34 - 35 | 4962 | 81 | |
AK001788.1 | - | 7 - 8 | 855 | 81 | |
AK022557.1 | - | 10 - 11 | 1310 | 81 |