Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAA4
Entrez Gene ID
6291
Gene Name
serum amyloid A4, constitutive
Gene Aliases
C-SAA, CSAA
Chromosome Location
Chr.11: 18231355 - 18236837 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006512.3 NP_006503.2 2 - 3 216 67
GenBank mRNA AK312182.1 - 2 - 3 167 67
AV699045.1 - 2 - 3 159 67
BC007026.1 - 2 - 3 148 67
BG569210.1 - 2 - 3 217 67
BU685219.1 - 2 - 3 67
CB113387.1 - 2 - 3 189 67
CR541758.1 - 1 - 2 91 67
M81349.1 - 2 - 3 166 67

Gene Symbol
SAA2-SAA4
Entrez Gene ID
100528017
Gene Name
SAA2-SAA4 readthrough
Gene Aliases
-
Chromosome Location
Chr.11: 18231355 - 18248674 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199744.1 NP_001186673.1 4 - 5 401 67
GenBank mRNA BG616641.1 - 4 - 5 363 67
BX117371.1 - 4 - 5 345 67

View Full Product Details