The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006532.3 | NP_006523.1 | 8 - 9 | 1540 | 95 |
XM_017027335.1 | XP_016882824.1 | 9 - 10 | 1524 | 95 | |
XM_017027336.1 | XP_016882825.1 | 4 - 5 | 1276 | 95 | |
XM_017027337.1 | XP_016882826.1 | 3 - 4 | 716 | 95 | |
GenBank mRNA | AB208794.1 | - | 9 - 10 | 1521 | 95 |
AK293105.1 | - | 8 - 9 | 1473 | 95 | |
BC049195.1 | - | 8 - 9 | 1522 | 95 | |
U16282.1 | - | 8 - 9 | 1480 | 95 |