The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006668.1 | NP_006659.1 | 5 - 6 | 446 | 70 |
XM_011536364.1 | XP_011534666.1 | 5 - 6 | 602 | 70 | |
XM_017020933.1 | XP_016876422.1 | 6 - 7 | 645 | 70 | |
GenBank mRNA | AB209749.1 | - | 6 - 7 | 603 | 70 |
AF094480.1 | - | 5 - 6 | 446 | 70 | |
AK295216.1 | - | 5 - 6 | 408 | 70 | |
BC022539.1 | - | 5 - 6 | 547 | 70 |