Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTMR11
Entrez Gene ID
10903
Gene Name
myotubularin related protein 11
Gene Aliases
CRA
Chromosome Location
Chr.1: 149928651 - 149936898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC032655.1 - 6 - 7 614 63
U78557.1 - 7 - 8 868 63

View Full Product Details