The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006699.3 | NP_006690.1 | 1 - 2 | 1023 | 80 |
XM_006710302.2 | XP_006710365.1 | 1 - 2 | 1031 | 80 | |
XM_011540536.2 | XP_011538838.1 | 1 - 2 | 1093 | 80 | |
XM_017000115.1 | XP_016855604.1 | 1 - 2 | 1093 | 80 | |
GenBank mRNA | AF027156.1 | - | 1 - 2 | 822 | 80 |
AK307842.1 | - | 1 - 2 | 1031 | 80 | |
BC063300.1 | - | 1 - 2 | 1021 | 80 |