Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJB2
Entrez Gene ID
3300
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
Gene Aliases
CMT2T, DSMA5, HSJ-1, HSJ1, HSPF3
Chromosome Location
Chr.2: 219279318 - 219286900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039550.1 NP_001034639.1 5 - 6 590 111
NM_006736.5 NP_006727.2 5 - 6 590 111
GenBank mRNA AK289626.1 - 5 - 6 413 111
AK292761.1 - 5 - 6 418 111
AK312723.1 - 5 - 6 597 111
AL547521.3 - 5 - 6 636 111
BC011609.2 - 5 - 6 532 111
BC047056.1 - 4 - 5 393 111
BT007088.1 - 4 - 5 353 111
BX420085.2 - - 592 111
S37374.1 - 4 - 5 378 111
S37375.1 - 4 - 5 378 111

View Full Product Details