Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAPTM5
Entrez Gene ID
7805
Gene Name
lysosomal protein transmembrane 5
Gene Aliases
CLAST6
Chromosome Location
Chr.1: 30732468 - 30757840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006762.2 NP_006753.1 3 - 4 347 67
GenBank mRNA AK292009.1 - 3 - 4 297 67
AK297014.1 - 3 - 4 285 67
BC106896.1 - 3 - 4 306 67
BC106897.1 - 3 - 4 306 67
CT003824.1 - 3 - 4 347 67
D42042.1 - 3 - 4 277 67
U30498.1 - 3 - 4 67
U51240.1 - 3 - 4 331 67

View Full Product Details