Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLIPR1
Entrez Gene ID
11010
Gene Name
GLI pathogenesis related 1
Gene Aliases
CRISP7, GLIPR, RTVP1
Chromosome Location
Chr.12: 75480733 - 75501936 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006851.2 NP_006842.2 3 - 4 680 86
XM_011537775.2 XP_011536077.1 4 - 5 730 86
XM_017018721.1 XP_016874210.1 3 - 4 560 86
GenBank mRNA AF400440.1 - 3 - 4 560 86
AK313449.1 - 3 - 4 546 86
BC012510.1 - 3 - 4 596 86
EF667354.1 - 4 - 5 603 86
U16307.3 - 3 - 4 616 86
X91911.1 - 3 - 4 660 86

View Full Product Details