The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006915.2 | NP_008846.2 | 2 - 3 | 960 | 89 |
GenBank mRNA | AJ007590.1 | - | 2 - 3 | 943 | 89 |
AK312555.1 | - | 2 - 3 | 794 | 89 | |
BC043348.2 | - | 2 - 3 | 882 | 89 | |
BC053530.1 | - | 2 - 3 | 880 | 89 |