Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RNF24
Entrez Gene ID
11237
Gene Name
ring finger protein 24
Gene Aliases
G1L
Chromosome Location
Chr.20: 3927309 - 4015591 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007219.4 NP_009150.1 1 - 2 451 71
GenBank mRNA AL096778.1 - 1 - 2 249 71

View Full Product Details