Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 166195271-166195774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929680
Entrez Gene ID
101929680
Gene Name
uncharacterized LOC101929680
Gene Aliases
-
UniGene
Hs.662210
Gene Chromosome Location
Chr.2: 166081531-166301784 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2

Gene Symbol
SCN9A
Entrez Gene ID
6335
Gene Name
sodium voltage-gated channel alpha subunit 9
Gene Aliases
ETHA, FEB3B, GEFSP7, HSAN2D, NE-NA, NENA, Nav1.7, PN1, SFNP
UniGene
Hs.439145
Gene Chromosome Location
Chr.2: 166195185-166375987 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2

SNP Detail

SNP ID Chromosome Location
rs1032836153 Chr.2: 166195600-166195600 on Build GRCh38
rs200598248 Chr.2: 166195745-166195745 on Build GRCh38
rs186613025 Chr.2: 166195658-166195658 on Build GRCh38
rs1048607018 Chr.2: 166195461-166195461 on Build GRCh38
rs955253702 Chr.2: 166195385-166195385 on Build GRCh38
rs899603476 Chr.2: 166195455-166195455 on Build GRCh38
rs202149808 Chr.2: 166195575-166195575 on Build GRCh38
rs965267449 Chr.2: 166195674-166195674 on Build GRCh38
rs1015184575 Chr.2: 166195470-166195470 on Build GRCh38
rs191667986 Chr.2: 166195355-166195355 on Build GRCh38
rs200582772 Chr.2: 166195585-166195585 on Build GRCh38
rs201419776 Chr.2: 166195309-166195309 on Build GRCh38
rs199744543 Chr.2: 166195303-166195303 on Build GRCh38
rs990286882 Chr.2: 166195415-166195415 on Build GRCh38
rs139483482 Chr.2: 166195390-166195390 on Build GRCh38
rs201730339 Chr.2: 166195686-166195686 on Build GRCh38
rs537634344 Chr.2: 166195604-166195604 on Build GRCh38
rs957534160 Chr.2: 166195599-166195599 on Build GRCh38
rs989969928 Chr.2: 166195610-166195610 on Build GRCh38
rs928983401 Chr.2: 166195682-166195682 on Build GRCh38
rs201300071 Chr.2: 166195468-166195468 on Build GRCh38
rs908767208 Chr.2: 166195701-166195701 on Build GRCh38
rs200841817 Chr.2: 166195513-166195513 on Build GRCh38
rs939578730 Chr.2: 166195493-166195493 on Build GRCh38
rs1045389187 Chr.2: 166195477-166195477 on Build GRCh38
rs1031609496 Chr.2: 166195591-166195591 on Build GRCh38
rs200630689 Chr.2: 166195659-166195659 on Build GRCh38
rs552197602 Chr.2: 166195383-166195383 on Build GRCh38
rs988347839 Chr.2: 166195644-166195644 on Build GRCh38
rs202150709 Chr.2: 166195565-166195565 on Build GRCh38
rs969420769 Chr.2: 166195344-166195344 on Build GRCh38
rs943695862 Chr.2: 166195433-166195433 on Build GRCh38
rs1029626458 Chr.2: 166195381-166195381 on Build GRCh38
rs534336578 Chr.2: 166195395-166195395 on Build GRCh38
rs202032498 Chr.2: 166195476-166195476 on Build GRCh38
rs200527016 Chr.2: 166195621-166195621 on Build GRCh38
rs1039359879 Chr.2: 166195447-166195447 on Build GRCh38
rs201694494 Chr.2: 166195586-166195586 on Build GRCh38
rs1017044114 Chr.2: 166195356-166195356 on Build GRCh38
rs899883386 Chr.2: 166195564-166195564 on Build GRCh38
rs200148433 Chr.2: 166195467-166195467 on Build GRCh38
rs887256984 Chr.2: 166195506-166195506 on Build GRCh38
rs201362859 Chr.2: 166195571-166195571 on Build GRCh38
rs753058763 Chr.2: 166195414-166195414 on Build GRCh38
rs199988048 Chr.2: 166195507-166195507 on Build GRCh38
rs199559478 Chr.2: 166195744-166195744 on Build GRCh38
rs182687583 Chr.2: 166195570-166195570 on Build GRCh38
rs199569173 Chr.2: 166195341-166195341 on Build GRCh38
rs200715030 Chr.2: 166195342-166195342 on Build GRCh38
rs935149231 Chr.2: 166195377-166195377 on Build GRCh38
rs201826787 Chr.2: 166195408-166195408 on Build GRCh38
rs199656729 Chr.2: 166195619-166195619 on Build GRCh38
rs1038958998 Chr.2: 166195515-166195515 on Build GRCh38
rs200910116 Chr.2: 166195305-166195305 on Build GRCh38
rs906868759 Chr.2: 166195488-166195488 on Build GRCh38
rs988111823 Chr.2: 166195378-166195378 on Build GRCh38
rs1047377228 Chr.2: 166195498-166195498 on Build GRCh38
rs201581806 Chr.2: 166195350-166195350 on Build GRCh38
rs374410124 Chr.2: 166195375-166195375 on Build GRCh38
rs896713230 Chr.2: 166195568-166195568 on Build GRCh38
rs941617888 Chr.2: 166195709-166195709 on Build GRCh38
rs931075076 Chr.2: 166195459-166195459 on Build GRCh38
rs115766730 Chr.2: 166195634-166195634 on Build GRCh38

View Full Product Details