Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 166196577-166197105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC101929680
Entrez Gene ID
101929680
Gene Name
uncharacterized LOC101929680
Gene Aliases
-
UniGene
Hs.662210
Gene Chromosome Location
Chr.2: 166081531-166301784 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2

Gene Symbol
SCN9A
Entrez Gene ID
6335
Gene Name
sodium voltage-gated channel alpha subunit 9
Gene Aliases
ETHA, FEB3B, GEFSP7, HSAN2D, NE-NA, NENA, Nav1.7, PN1, SFNP
UniGene
Hs.439145
Gene Chromosome Location
Chr.2: 166195185-166375987 on Build GRCh38
Transcripts
NR_110260.1 NM_002977.3 XM_005246757.2

SNP Detail

SNP ID Chromosome Location
rs201539480 Chr.2: 166196657-166196657 on Build GRCh38
rs969758416 Chr.2: 166196732-166196732 on Build GRCh38
rs16851751 Chr.2: 166196876-166196876 on Build GRCh38
rs200049536 Chr.2: 166196767-166196767 on Build GRCh38
rs201967308 Chr.2: 166196950-166196950 on Build GRCh38
rs1019846579 Chr.2: 166196863-166196863 on Build GRCh38
rs199789932 Chr.2: 166196635-166196635 on Build GRCh38
rs1037898062 Chr.2: 166196984-166196984 on Build GRCh38
rs1057174896 Chr.2: 166196928-166196928 on Build GRCh38
rs202141634 Chr.2: 166196899-166196899 on Build GRCh38
rs201050175 Chr.2: 166197060-166197060 on Build GRCh38
rs567068872 Chr.2: 166196991-166196991 on Build GRCh38
rs995940408 Chr.2: 166196849-166196849 on Build GRCh38
rs1037361780 Chr.2: 166196994-166196994 on Build GRCh38
rs938572129 Chr.2: 166196734-166196734 on Build GRCh38
rs992417935 Chr.2: 166196714-166196714 on Build GRCh38
rs868363066 Chr.2: 166196610-166196610 on Build GRCh38
rs995871381 Chr.2: 166196992-166196992 on Build GRCh38
rs907756721 Chr.2: 166196965-166196965 on Build GRCh38
rs201353462 Chr.2: 166196809-166196809 on Build GRCh38
rs202073550 Chr.2: 166196709-166196710 on Build GRCh38
rs201350089 Chr.2: 166196911-166196911 on Build GRCh38
rs901155232 Chr.2: 166196867-166196867 on Build GRCh38
rs199848927 Chr.2: 166197067-166197067 on Build GRCh38
rs1005101277 Chr.2: 166196769-166196769 on Build GRCh38
rs1008494092 Chr.2: 166197049-166197049 on Build GRCh38
rs200628780 Chr.2: 166197073-166197073 on Build GRCh38
rs979803899 Chr.2: 166196739-166196739 on Build GRCh38
rs768732014 Chr.2: 166196982-166196982 on Build GRCh38
rs938689853 Chr.2: 166196919-166196919 on Build GRCh38
rs1041042907 Chr.2: 166196866-166196866 on Build GRCh38
rs530429883 Chr.2: 166196716-166196716 on Build GRCh38
rs148459420 Chr.2: 166197067-166197067 on Build GRCh38
rs890101185 Chr.2: 166197022-166197022 on Build GRCh38
rs200200620 Chr.2: 166196830-166196830 on Build GRCh38
rs936847369 Chr.2: 166196753-166196753 on Build GRCh38
rs935649851 Chr.2: 166196993-166196993 on Build GRCh38
rs200644493 Chr.2: 166196745-166196745 on Build GRCh38
rs200614076 Chr.2: 166196962-166196962 on Build GRCh38
rs1008420991 Chr.2: 166196858-166196858 on Build GRCh38
rs1013896563 Chr.2: 166197032-166197032 on Build GRCh38
rs201537816 Chr.2: 166196976-166196976 on Build GRCh38
rs200091138 Chr.2: 166197066-166197067 on Build GRCh38
rs539504178 Chr.2: 166196880-166196880 on Build GRCh38
rs199684109 Chr.2: 166196985-166196985 on Build GRCh38
rs199551913 Chr.2: 166196712-166196712 on Build GRCh38
rs200353065 Chr.2: 166196902-166196902 on Build GRCh38
rs201582211 Chr.2: 166196748-166196748 on Build GRCh38
rs990180287 Chr.2: 166196754-166196754 on Build GRCh38
rs202203220 Chr.2: 166196857-166196857 on Build GRCh38
rs201173517 Chr.2: 166196831-166196831 on Build GRCh38
rs914749816 Chr.2: 166196889-166196889 on Build GRCh38
rs114843828 Chr.2: 166196898-166196898 on Build GRCh38
rs200781014 Chr.2: 166196637-166196637 on Build GRCh38
rs200558948 Chr.2: 166196795-166196795 on Build GRCh38
rs1028611299 Chr.2: 166196854-166196854 on Build GRCh38
rs1015506917 Chr.2: 166196638-166196638 on Build GRCh38
rs775434756 Chr.2: 166196894-166196894 on Build GRCh38
rs16851753 Chr.2: 166197012-166197012 on Build GRCh38
rs940624727 Chr.2: 166196981-166196981 on Build GRCh38
rs561611241 Chr.2: 166196660-166196660 on Build GRCh38
rs190442246 Chr.2: 166197065-166197065 on Build GRCh38

View Full Product Details