The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109974.2 | NP_001103444.1 | 1 - 2 | 210 | 85 |
NM_007286.5 | NP_009217.3 | 1 - 2 | 210 | 85 | |
GenBank mRNA | AB028952.1 | - | 1 - 2 | 164 | 85 |
AF499136.1 | - | 1 - 2 | 124 | 85 | |
AF499137.1 | - | 1 - 2 | 124 | 85 | |
AK290178.1 | - | 1 - 2 | 193 | 85 | |
BC142683.1 | - | 1 - 2 | 131 | 85 | |
BC146665.1 | - | 1 - 2 | 177 | 85 | |
BC152538.1 | - | 1 - 2 | 164 | 85 | |
BP237193.1 | - | 1 - 2 | 197 | 85 | |
DA333058.1 | - | 1 - 2 | 211 | 85 |