Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLEC1
Entrez Gene ID
9940
Gene Name
deleted in lung and esophageal cancer 1
Gene Aliases
CFAP81, DLC-1, DLC1, F56
Chromosome Location
Chr.3: 38038595 - 38122741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321153.1 NP_001308082.1 33 - 34 4738 82
NM_007335.3 NP_031361.2 33 - 34 4729 82
NM_007337.3 NP_031363.2 33 - 34 4729 82
XM_011534317.2 XP_011532619.1 33 - 34 5346 82
XM_011534318.2 XP_011532620.1 32 - 33 5173 82
GenBank mRNA AB020522.1 - 33 - 34 4729 82
AB425198.1 - 33 - 34 4710 82
AL137282.1 - 2 - 3 792 82
AL137706.1 - 21 - 22 3611 82
BC171799.1 - 33 - 34 4729 82

View Full Product Details