Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATRN
Entrez Gene ID
8455
Gene Name
attractin
Gene Aliases
DPPT-L, MGCA
Chromosome Location
Chr.20: 3471018 - 3651122 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207047.2 NP_001193976.1 15 - 16 2252 98
NM_001323332.1 NP_001310261.1 15 - 16 2635 98
NM_139321.2 NP_647537.1 15 - 16 2635 98
NM_139322.3 NP_647538.1 15 - 16 2635 98
GenBank mRNA AF106861.1 - 15 - 16 2545 98
AK293010.1 - 15 - 16 2635 98
AK302730.1 - 15 - 16 2231 98
BC101705.1 - 15 - 16 2585 98

View Full Product Details