The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012121.4 | NP_036253.2 | 1 - 2 | 112 | 150 |
GenBank mRNA | AK027385.1 | - | 1 - 2 | 103 | 150 |
AK097835.1 | - | 1 - 2 | 101 | 150 | |
AK312536.1 | - | 1 - 2 | 102 | 150 | |
BC002774.1 | - | 1 - 2 | 112 | 150 | |
BE387060.1 | - | 1 - 2 | 111 | 150 | |
BI560730.1 | - | 1 - 2 | 135 | 150 |