The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012138.3 | NP_036270.1 | 8 - 9 | 1653 | 73 |
XM_011524611.1 | XP_011522913.1 | 8 - 9 | 1637 | 73 | |
GenBank mRNA | AF083208.1 | - | 8 - 9 | 1567 | 73 |
AF161395.1 | - | 3 - 4 | 323 | 73 | |
AJ249940.2 | - | 8 - 9 | 1508 | 73 | |
AK026173.1 | - | 6 - 7 | 1079 | 73 | |
AK057229.1 | - | 8 - 9 | 1653 | 73 | |
BC000591.2 | - | 8 - 9 | 1533 | 73 |