The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012164.3 | NP_036296.2 | 2 - 3 | 217 | 108 |
XM_005251910.3 | XP_005251967.1 | 2 - 3 | 220 | 108 | |
XM_017014615.1 | XP_016870104.1 | 2 - 3 | 220 | 108 | |
GenBank mRNA | AF145024.1 | - | 2 - 3 | 124 | 108 |
AF176698.1 | - | 1 - 2 | 102 | 108 | |
AF193594.1 | - | 2 - 3 | 124 | 108 | |
AK022484.1 | - | 2 - 3 | 167 | 108 | |
BC018738.1 | - | 2 - 3 | 133 | 108 | |
BC110334.1 | - | 1 - 2 | 117 | 108 |