The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033583.2 | NP_001028755.2 | 8 - 9 | 783 | 60 |
NM_001037171.1 | NP_001032248.1 | 9 - 10 | 810 | 60 | |
XM_005274472.2 | XP_005274529.1 | 7 - 8 | 682 | 60 | |
GenBank mRNA | AF132950.1 | - | 7 - 8 | 685 | 60 |
AK000659.1 | - | 8 - 9 | 769 | 60 | |
AK024337.1 | - | 10 - 11 | 1875 | 60 | |
BC136671.1 | - | 8 - 9 | 734 | 60 | |
BC144360.1 | - | 8 - 9 | 734 | 60 | |
BX648512.1 | - | 9 - 10 | 794 | 60 |