Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAGLN3
Entrez Gene ID
29114
Gene Name
transgelin 3
Gene Aliases
NP22, NP24, NP25
Chromosome Location
Chr.3: 111998739 - 112013888 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008272.1 NP_001008273.1 2 - 3 557 84
NM_001008273.1 NP_001008274.1 1 - 2 442 84
NM_013259.2 NP_037391.2 2 - 3 731 84
GenBank mRNA AF087855.1 - 2 - 3 443 84
AF112201.1 - 1 - 2 228 84
AF303058.1 - 2 - 3 596 84
AK315289.1 - 2 - 3 415 84
BC015329.1 - 2 - 3 313 84
BI752099.1 - 2 - 3 632 84
BI822856.1 - 1 - 2 378 84
BT007408.1 - 1 - 2 179 84
CK003985.1 - 1 - 2 239 84

View Full Product Details