The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013385.4 | NP_037517.1 | 1 - 2 | 206 | 86 |
XM_011530148.1 | XP_011528450.1 | 1 - 2 | 55 | 86 | |
GenBank mRNA | AF075458.1 | - | 1 - 2 | 135 | 86 |
AF125349.1 | - | 1 - 2 | 206 | 86 | |
AK024428.1 | - | 1 - 2 | 60 | 86 | |
AK315052.1 | - | 1 - 2 | 42 | 86 | |
BC017780.1 | - | 1 - 2 | 74 | 86 | |
BC041161.1 | - | 1 - 2 | 64 | 86 | |
BP306826.1 | - | 1 - 2 | 99 | 86 | |
CR456551.1 | - | 1 - 2 | 55 | 86 |