The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014037.2 | NP_054756.2 | 7 - 8 | 1464 | 78 |
XM_005258820.2 | XP_005258877.1 | 8 - 9 | 1565 | 78 | |
XM_006723168.3 | XP_006723231.1 | 7 - 8 | 1480 | 78 | |
XM_011526859.2 | XP_011525161.2 | 6 - 7 | 1387 | 78 | |
XM_011526860.2 | XP_011525162.1 | 8 - 9 | 1777 | 78 | |
XM_011526861.2 | XP_011525163.1 | 6 - 7 | 887 | 78 | |
XM_017026712.1 | XP_016882201.1 | 7 - 8 | 1480 | 78 | |
GenBank mRNA | AF151977.1 | - | 6 - 7 | 1272 | 78 |
AF265578.1 | - | 6 - 7 | 1230 | 78 | |
AL136856.1 | - | 6 - 7 | 1276 | 78 | |
BC034948.1 | - | 7 - 8 | 1432 | 78 |