Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSORS1C1
Entrez Gene ID
170679
Gene Name
psoriasis susceptibility 1 candidate 1
Gene Aliases
C6orf16, SEEK1
Chromosome Location
Chr.6: 31114831 - 31140092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014068.2 NP_054787.2 2 - 3 226 90
GenBank mRNA AB031479.1 - 2 - 3 211 90
AF484418.1 - 2 - 3 227 90

View Full Product Details