The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014169.3 | NP_054888.2 | 1 - 2 | 421 | 78 |
GenBank mRNA | AB100262.2 | - | 1 - 2 | 56 | 78 |
AF161483.1 | - | 1 - 2 | 75 | 78 | |
AF212243.1 | - | 1 - 2 | 51 | 78 | |
AY329084.1 | - | 1 - 2 | 30 | 78 | |
BC010893.1 | - | 1 - 2 | 58 | 78 | |
BC107699.1 | - | 1 - 2 | 83 | 78 | |
BC113533.1 | - | 1 - 2 | 421 | 78 | |
BC113535.1 | - | 1 - 2 | 421 | 78 |