The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014170.2 | NP_054889.2 | 5 - 6 | 834 | 63 |
NM_138485.1 | NP_612494.1 | 4 - 5 | 735 | 63 | |
GenBank mRNA | AF161484.1 | - | 5 - 6 | 873 | 63 |
AK289611.1 | - | 4 - 5 | 697 | 63 | |
AK292651.1 | - | 5 - 6 | 853 | 63 | |
BC010137.2 | - | 3 - 4 | 570 | 63 | |
BC037163.1 | - | 5 - 6 | 837 | 63 | |
BX354218.2 | - | 4 - 5 | 703 | 63 | |
CA429378.1 | - | 1 - 2 | 63 |