The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014182.4 | NP_054901.1 | 1 - 2 | 89 | 96 |
GenBank mRNA | AF161509.1 | - | 1 - 2 | 50 | 96 |
AF173375.1 | - | 1 - 2 | 59 | 96 | |
AF395707.1 | - | 1 - 2 | 186 | 96 | |
AK314046.1 | - | 1 - 2 | 70 | 96 |