The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014306.4 | NP_055121.1 | 11 - 12 | 1540 | 102 |
GenBank mRNA | AF155658.1 | - | 11 - 12 | 1488 | 102 |
AF161466.1 | - | 11 - 12 | 1499 | 102 | |
AK297908.1 | - | - | 698 | 102 | |
AK312503.1 | - | 11 - 12 | 1500 | 102 | |
AL050255.1 | - | 11 - 12 | 1484 | 102 | |
AL137272.1 | - | 4 - 5 | 486 | 102 | |
BC000151.1 | - | 11 - 12 | 1478 | 102 | |
BC002970.1 | - | 11 - 12 | 1478 | 102 | |
BC010308.1 | - | 11 - 12 | 1490 | 102 | |
BC016707.1 | - | 11 - 12 | 1505 | 102 | |
BU615556.1 | - | 1 - 2 | 102 | ||
CR456450.1 | - | 11 - 12 | 1455 | 102 | |
CR457175.1 | - | 11 - 12 | 1409 | 102 |