The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164380.1 | NP_001157852.1 | 1 - 2 | 145 | 111 |
NM_001164381.1 | NP_001157853.1 | 1 - 2 | 145 | 111 | |
NM_001164382.1 | NP_001157854.1 | 1 - 2 | 145 | 111 | |
NM_014393.2 | NP_055208.2 | 1 - 2 | 145 | 111 | |
GenBank mRNA | AB210014.1 | - | 1 - 2 | 41 | 111 |
AK002152.1 | - | 1 - 2 | 58 | 111 | |
AK303104.1 | - | 1 - 2 | 58 | 111 | |
AK314617.1 | - | 1 - 2 | 58 | 111 | |
BC008369.1 | - | 1 - 2 | 58 | 111 | |
BC008370.1 | - | 1 - 2 | 58 | 111 | |
Y19062.1 | - | 1 - 2 | 40 | 111 |