The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014445.3 | NP_055260.1 | 2 - 3 | 665 | 79 |
XM_011512698.2 | XP_011511000.1 | 3 - 4 | 1330 | 79 | |
GenBank mRNA | AB022427.1 | - | 2 - 3 | 373 | 79 |
AF136975.1 | - | 2 - 3 | 528 | 79 | |
AK090933.1 | - | 3 - 4 | 477 | 79 | |
AK093929.1 | - | 3 - 4 | 486 | 79 | |
AK098135.1 | - | 3 - 4 | 381 | 79 | |
AK125413.1 | - | 3 - 4 | 1431 | 79 | |
AL136807.1 | - | 2 - 3 | 474 | 79 | |
BC108314.1 | - | 2 - 3 | 513 | 79 | |
BC112364.1 | - | 2 - 3 | 472 | 79 | |
BC112365.1 | - | 2 - 3 | 472 | 79 | |
CR533472.1 | - | 2 - 3 | 159 | 79 | |
X75694.1 | - | 2 - 3 | 142 | 79 |