The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014475.3 | NP_055290.1 | 4 - 5 | 660 | 77 |
XM_005258748.4 | XP_005258805.1 | 3 - 4 | 1057 | 77 | |
XM_017026598.1 | XP_016882087.1 | 4 - 5 | 1220 | 77 | |
GenBank mRNA | AB021933.1 | - | 4 - 5 | 620 | 77 |
BC032730.1 | - | 4 - 5 | 671 | 77 |