The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014482.1 | NP_055297.1 | 1 - 2 | 500 | 71 |
GenBank mRNA | AF101441.1 | - | 1 - 2 | 500 | 71 |
BC069080.1 | - | 1 - 2 | 354 | 71 | |
BC101734.1 | - | 1 - 2 | 364 | 71 | |
BC105063.1 | - | 1 - 2 | 364 | 71 |