The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003793.2 | NP_001003793.1 | 1 - 2 | 447 | 105 |
NM_001177711.1 | NP_001171182.1 | 1 - 2 | 447 | 105 | |
NM_001177712.1 | NP_001171183.1 | 1 - 2 | 447 | 105 | |
NM_014483.3 | NP_055298.2 | 1 - 2 | 447 | 105 | |
XM_005265061.2 | XP_005265118.1 | 1 - 2 | 899 | 105 | |
XM_005265062.1 | XP_005265119.1 | 1 - 2 | 899 | 105 | |
XM_017006180.1 | XP_016861669.1 | 1 - 2 | 899 | 105 | |
GenBank mRNA | AF023259.1 | - | 1 - 2 | 224 | 105 |
AK292789.1 | - | 1 - 2 | 123 | 105 | |
AY236871.1 | - | 1 - 2 | 307 | 105 | |
BC117315.1 | - | 1 - 2 | 144 | 105 | |
BC143521.1 | - | 1 - 2 | 144 | 105 |