The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014502.4 | NP_055317.1 | 1 - 2 | 225 | 98 |
GenBank mRNA | AF086469.1 | - | 1 - 2 | 175 | 98 |
AJ131186.1 | - | 1 - 2 | 225 | 98 | |
AL555169.3 | - | 1 - 2 | 235 | 99 | |
BC008719.2 | - | 1 - 2 | 220 | 98 | |
BC018665.2 | - | 1 - 2 | 220 | 98 | |
BC018698.2 | - | 1 - 2 | 220 | 98 |