Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXC4
Entrez Gene ID
3221
Gene Name
homeobox C4
Gene Aliases
HOX3, HOX3E, cp19
Chromosome Location
Chr.12: 54016852 - 54056030 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014620.5 NP_055435.2 2 - 3 673 104
GenBank mRNA X07495.1 - 2 - 3 594 104

View Full Product Details