The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303012.1 | NP_001289941.1 | 19 - 20 | 2663 | 70 |
NM_001303013.1 | NP_001289942.1 | 19 - 20 | 3849 | 70 | |
NM_014638.3 | NP_055453.2 | 19 - 20 | 2857 | 70 | |
XM_011542455.2 | XP_011540757.1 | 20 - 21 | 2899 | 70 | |
XM_011542456.2 | XP_011540758.1 | 20 - 21 | 2856 | 70 | |
XM_017002870.1 | XP_016858359.1 | 20 - 21 | 2888 | 70 | |
XM_017002871.1 | XP_016858360.1 | 20 - 21 | 2859 | 70 | |
XM_017002872.1 | XP_016858361.1 | 20 - 21 | 2888 | 70 | |
XM_017002873.1 | XP_016858362.1 | 19 - 20 | 2780 | 70 | |
XM_017002874.1 | XP_016858363.1 | 20 - 21 | 2888 | 70 | |
GenBank mRNA | AB007919.2 | - | 19 - 20 | 2663 | 70 |
AK074149.1 | - | 8 - 9 | 919 | 70 | |
AK122589.1 | - | 17 - 18 | 2956 | 70 | |
AK122591.2 | - | 6 - 7 | 802 | 70 | |
BC019679.1 | - | 5 - 6 | 485 | 70 | |
BC043358.1 | - | 16 - 17 | 2051 | 70 | |
BC044630.1 | - | 2 - 3 | 211 | 70 | |
BC128207.1 | - | 17 - 18 | 2239 | 70 | |
BC144620.1 | - | 19 - 20 | 3841 | 70 | |
DQ176850.1 | - | 19 - 20 | 2857 | 70 |