Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218274506-218275012 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNKD
Entrez Gene ID
25953
Gene Name
paroxysmal nonkinesigenic dyskinesia
Gene Aliases
BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2
UniGene
Hs.98475
Gene Chromosome Location
Chr.2: 218270392-218346793 on Build GRCh38
Transcripts
NM_015488.4 XM_017003771.1 NM_001321427.1 NM_001321428.1 NM_001321429.1 NM_001321430.1 NM_001321432.1 NM_001321433.1 NM_001321435.1 NM_001321436.1 NM_001321438.1 NM_022152.5 NR_135643.1 XM_011511627.1

Gene Symbol
TMBIM1
Entrez Gene ID
64114
Gene Name
transmembrane BAX inhibitor motif containing 1
Gene Aliases
LFG3, MST100, MSTP100, PP1201, RECS1
UniGene
Hs.591605
Gene Chromosome Location
Chr.2: 218274192-218292577 on Build GRCh38
Transcripts
NM_015488.4 XM_017003771.1 NM_001321427.1 NM_001321428.1 NM_001321429.1 NM_001321430.1 NM_001321432.1 NM_001321433.1 NM_001321435.1 NM_001321436.1 NM_001321438.1 NM_022152.5 NR_135643.1 XM_011511627.1

SNP Detail

SNP ID Chromosome Location
rs539236010 Chr.2: 218274844-218274844 on Build GRCh38
rs964534483 Chr.2: 218274801-218274801 on Build GRCh38
rs751053599 Chr.2: 218274692-218274692 on Build GRCh38
rs1018754282 Chr.2: 218274583-218274583 on Build GRCh38
rs1003596204 Chr.2: 218274881-218274881 on Build GRCh38
rs990835553 Chr.2: 218274973-218274973 on Build GRCh38
rs748478628 Chr.2: 218274910-218274910 on Build GRCh38
rs951904519 Chr.2: 218274845-218274845 on Build GRCh38
rs868124955 Chr.2: 218274586-218274586 on Build GRCh38
rs898436292 Chr.2: 218274775-218274775 on Build GRCh38
rs11552007 Chr.2: 218274765-218274765 on Build GRCh38
rs907878707 Chr.2: 218274668-218274668 on Build GRCh38
rs1008549690 Chr.2: 218274772-218274772 on Build GRCh38
rs950888169 Chr.2: 218274536-218274536 on Build GRCh38
rs1035108115 Chr.2: 218274884-218274884 on Build GRCh38
rs765721734 Chr.2: 218274683-218274683 on Build GRCh38
rs538459642 Chr.2: 218274588-218274588 on Build GRCh38
rs569581018 Chr.2: 218274750-218274750 on Build GRCh38
rs201491578 Chr.2: 218274707-218274707 on Build GRCh38
rs116809757 Chr.2: 218274757-218274757 on Build GRCh38
rs907846067 Chr.2: 218274955-218274955 on Build GRCh38
rs983416971 Chr.2: 218274647-218274647 on Build GRCh38
rs550508792 Chr.2: 218274817-218274817 on Build GRCh38
rs571839302 Chr.2: 218274825-218274825 on Build GRCh38
rs959338710 Chr.2: 218274933-218274933 on Build GRCh38
rs946878485 Chr.2: 218274700-218274700 on Build GRCh38
rs950089135 Chr.2: 218274675-218274675 on Build GRCh38
rs987202085 Chr.2: 218274575-218274575 on Build GRCh38
rs536393381 Chr.2: 218274878-218274878 on Build GRCh38
rs933767860 Chr.2: 218274709-218274709 on Build GRCh38
rs559032091 Chr.2: 218274535-218274535 on Build GRCh38
rs755286764 Chr.2: 218274765-218274768 on Build GRCh38
rs575767964 Chr.2: 218274859-218274859 on Build GRCh38
rs186968810 Chr.2: 218274539-218274539 on Build GRCh38
rs756484708 Chr.2: 218274987-218274987 on Build GRCh38
rs930582295 Chr.2: 218274616-218274616 on Build GRCh38
rs1038222374 Chr.2: 218274773-218274773 on Build GRCh38
rs905975507 Chr.2: 218274690-218274690 on Build GRCh38
rs573048586 Chr.2: 218274862-218274862 on Build GRCh38
rs1050808789 Chr.2: 218274744-218274744 on Build GRCh38
rs915499515 Chr.2: 218274679-218274679 on Build GRCh38
rs2271542 Chr.2: 218274587-218274587 on Build GRCh38
rs1042970941 Chr.2: 218274706-218274706 on Build GRCh38
rs991032138 Chr.2: 218274655-218274655 on Build GRCh38
rs966396039 Chr.2: 218274537-218274537 on Build GRCh38
rs554927358 Chr.2: 218274891-218274891 on Build GRCh38
rs754200309 Chr.2: 218274555-218274555 on Build GRCh38
rs751710286 Chr.2: 218274622-218274622 on Build GRCh38
rs993997918 Chr.2: 218274778-218274778 on Build GRCh38
rs893369349 Chr.2: 218274711-218274711 on Build GRCh38
rs6734242 Chr.2: 218274850-218274850 on Build GRCh38

View Full Product Details