Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218274908-218275414 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNKD
Entrez Gene ID
25953
Gene Name
paroxysmal nonkinesigenic dyskinesia
Gene Aliases
BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2
UniGene
Hs.98475
Gene Chromosome Location
Chr.2: 218270392-218346793 on Build GRCh38
Transcripts
NM_015488.4 XM_017003771.1 NM_001321427.1 NM_001321428.1 NM_001321429.1 NM_001321430.1 NM_001321432.1 NM_001321433.1 NM_001321435.1 NM_001321436.1 NM_001321438.1 NM_022152.5 NR_135643.1 XM_011511627.1

Gene Symbol
TMBIM1
Entrez Gene ID
64114
Gene Name
transmembrane BAX inhibitor motif containing 1
Gene Aliases
LFG3, MST100, MSTP100, PP1201, RECS1
UniGene
Hs.591605
Gene Chromosome Location
Chr.2: 218274192-218292577 on Build GRCh38
Transcripts
NM_015488.4 XM_017003771.1 NM_001321427.1 NM_001321428.1 NM_001321429.1 NM_001321430.1 NM_001321432.1 NM_001321433.1 NM_001321435.1 NM_001321436.1 NM_001321438.1 NM_022152.5 NR_135643.1 XM_011511627.1

SNP Detail

SNP ID Chromosome Location
rs533937187 Chr.2: 218275006-218275006 on Build GRCh38
rs959338710 Chr.2: 218274933-218274933 on Build GRCh38
rs1054994825 Chr.2: 218275048-218275048 on Build GRCh38
rs924234188 Chr.2: 218275376-218275376 on Build GRCh38
rs367809004 Chr.2: 218275144-218275144 on Build GRCh38
rs755813383 Chr.2: 218274999-218274999 on Build GRCh38
rs1012248595 Chr.2: 218275261-218275261 on Build GRCh38
rs1041909648 Chr.2: 218275344-218275344 on Build GRCh38
rs994526173 Chr.2: 218275173-218275173 on Build GRCh38
rs990835553 Chr.2: 218274973-218274973 on Build GRCh38
rs924069461 Chr.2: 218275064-218275064 on Build GRCh38
rs182481983 Chr.2: 218275219-218275219 on Build GRCh38
rs959475135 Chr.2: 218275251-218275251 on Build GRCh38
rs563268273 Chr.2: 218275104-218275104 on Build GRCh38
rs544767253 Chr.2: 218275071-218275071 on Build GRCh38
rs906992379 Chr.2: 218275217-218275217 on Build GRCh38
rs981839942 Chr.2: 218275279-218275279 on Build GRCh38
rs12612648 Chr.2: 218275292-218275292 on Build GRCh38
rs756484708 Chr.2: 218274987-218274987 on Build GRCh38
rs745666436 Chr.2: 218275323-218275324 on Build GRCh38
rs1027877777 Chr.2: 218275229-218275229 on Build GRCh38
rs1035077028 Chr.2: 218275247-218275247 on Build GRCh38
rs978274819 Chr.2: 218275015-218275015 on Build GRCh38
rs1004747214 Chr.2: 218275257-218275257 on Build GRCh38
rs777776261 Chr.2: 218275157-218275157 on Build GRCh38
rs371347056 Chr.2: 218275079-218275079 on Build GRCh38
rs990267392 Chr.2: 218275300-218275300 on Build GRCh38
rs190669619 Chr.2: 218275051-218275051 on Build GRCh38
rs898410529 Chr.2: 218275169-218275169 on Build GRCh38
rs778856282 Chr.2: 218275388-218275388 on Build GRCh38
rs927365504 Chr.2: 218275041-218275041 on Build GRCh38
rs749023782 Chr.2: 218275291-218275291 on Build GRCh38
rs1040075021 Chr.2: 218275124-218275124 on Build GRCh38
rs937473643 Chr.2: 218275045-218275048 on Build GRCh38
rs1008517046 Chr.2: 218275112-218275112 on Build GRCh38
rs978708495 Chr.2: 218275336-218275336 on Build GRCh38
rs946201922 Chr.2: 218275343-218275343 on Build GRCh38
rs559333749 Chr.2: 218275232-218275232 on Build GRCh38
rs1002727986 Chr.2: 218275220-218275220 on Build GRCh38
rs907846067 Chr.2: 218274955-218274955 on Build GRCh38
rs529777981 Chr.2: 218275278-218275278 on Build GRCh38
rs1046983871 Chr.2: 218275215-218275215 on Build GRCh38
rs60211154 Chr.2: 218275103-218275103 on Build GRCh38
rs36035722 Chr.2: 218275177-218275178 on Build GRCh38

View Full Product Details