The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171904.1 | NP_001165375.1 | 9 - 10 | 1421 | 100 |
NM_014703.2 | NP_055518.1 | 10 - 11 | 1471 | 100 | |
XM_005276753.4 | XP_005276810.1 | 8 - 9 | 1302 | 100 | |
XM_005276755.4 | XP_005276812.1 | 9 - 10 | 1383 | 100 | |
XM_011534273.2 | XP_011532575.1 | 11 - 12 | 1562 | 100 | |
XM_011534274.2 | XP_011532576.1 | 11 - 12 | 1622 | 100 | |
XM_011534275.2 | XP_011532577.1 | 11 - 12 | 1537 | 100 | |
XM_011534276.2 | XP_011532578.1 | 11 - 12 | 1584 | 100 | |
XM_011534277.2 | XP_011532579.1 | 9 - 10 | 1455 | 100 | |
XM_017007546.1 | XP_016863035.1 | 10 - 11 | 1480 | 100 | |
XM_017007547.1 | XP_016863036.1 | 10 - 11 | 1437 | 100 | |
XM_017007548.1 | XP_016863037.1 | 10 - 11 | 1572 | 100 | |
XM_017007549.1 | XP_016863038.1 | 11 - 12 | 1622 | 100 | |
XM_017007550.1 | XP_016863039.1 | 11 - 12 | 1622 | 100 | |
XM_017007551.1 | XP_016863040.1 | 8 - 9 | 1332 | 100 | |
GenBank mRNA | AB018343.1 | - | 10 - 11 | 1454 | 100 |
AF061935.1 | - | 5 - 6 | 969 | 100 | |
AL080145.1 | - | 1 - 2 | 166 | 100 | |
BC110371.1 | - | 9 - 10 | 1411 | 100 |