The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014736.5 | NP_055551.1 | 3 - 4 | 430 | 60 |
NR_109934.1 | 3 - 4 | 486 | 60 | ||
GenBank mRNA | AF529370.1 | - | 3 - 4 | 291 | 60 |
AK290748.1 | - | 3 - 4 | 361 | 60 | |
AY598324.1 | - | 3 - 4 | 291 | 60 | |
BC005832.2 | - | 3 - 4 | 350 | 60 | |
BC007101.1 | - | 3 - 4 | 357 | 60 | |
BC016782.1 | - | 3 - 4 | 365 | 60 | |
BF691807.1 | - | 3 - 4 | 431 | 60 | |
BG502841.1 | - | - | 423 | 60 | |
D14657.1 | - | 3 - 4 | 352 | 60 | |
DT220867.1 | - | 3 - 4 | 483 | 60 |