Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAML1
Entrez Gene ID
9794
Gene Name
mastermind like transcriptional coactivator 1
Gene Aliases
Mam-1, Mam1
Chromosome Location
Chr.5: 179732850 - 179777286 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014757.4 NP_055572.1 2 - 3 1991 63
GenBank mRNA AB209135.1 - 3 - 4 1853 63
AF221759.1 - 1 - 2 1359 63
D83785.1 - 2 - 3 1991 63

View Full Product Details