Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
URB2
Entrez Gene ID
9816
Gene Name
URB2 ribosome biogenesis 2 homolog (S. cerevisiae)
Gene Aliases
KIAA0133, NET10, NPA2
Chromosome Location
Chr.1: 229626216 - 229660200 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314021.1 NP_001300950.1 5 - 6 3982 102
NM_014777.3 NP_055592.2 5 - 6 3951 102
GenBank mRNA AK291013.1 - 5 - 6 3951 102
BC114559.1 - 5 - 6 3904 102
BC114963.1 - 5 - 6 3873 102
D50923.2 - 5 - 6 3933 102

View Full Product Details